Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-iodopiperidine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43727-0010 43727-0050
TS43727-0010EA 104 USD InStock
TS43727-0010 TS43727-0050
1-Boc-4-iodopiperidine 97%
1-Boc-4-iodopiperidine


Formula: C₁₀H₁₈INO₂
Melting Pt: 44…49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115041
CAS Number: 301673-14-3
Order Now
Specification Test Results
Appearance White to light brown Crystalline powder or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty