Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfisoxazole 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43967-0250 43967-1000
TS43967-0250EA 45 USD InStock
TS43967-0250 TS43967-1000
Sulfisoxazole 99%
Sulfisoxazole


Formula: C₁₁H₁₃N₃O₃S
Melting Pt: 194 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003150
CAS Number: 127-69-5
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty