Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridine 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43994-0050 43994-0250
TS43994-0050EA 87 USD InStock
TS43994-0050 TS43994-0250
Imidazo[1,2-a]pyridine 99%
Imidazo[1,2-a]pyridine


Formula: C₇H₆N₂
MW: 118.1359 g/mol
Boiling Pt: 97…98 °C (760 mmHg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005553
CAS Number: 274-76-0
Order Now
Specification Test Results
Appearance Colorless to yellow to brown Liquid
Infrared spectrum Conforms
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty