Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44052-0050 44052-0250 44052-1000
TS44052-0050EA 34 USD InStock
TS44052-0050 TS44052-0250 TS44052-1000
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride


Formula: C₁₃H₁₆ClNOS
MW: 269.79 g/mol
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00011959
CAS Number: 4568-71-2
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 147 °C
Elemental Analysis 5.06 to 5.32 % N

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty