Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-pyrazinecarboxylate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44317-0010 44317-0050
TS44317-0010EA 28 USD InStock
TS44317-0010 TS44317-0050
Methyl-2-pyrazinecarboxylate 97%
Methyl-2-pyrazinecarboxylate


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 59.5…61.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00014611
CAS Number: 6164-79-0
Order Now
Specification Test Results
Appearance White to yellow to light orange Crystalline powder
Infrared spectrum Conforms
Melting point 57 to 63 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty