Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-Hydroxypyrrolidin-2-one 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44382-0010 44382-0050
TS44382-0010EA 133 USD InStock
TS44382-0010 TS44382-0050
(R)-4-Hydroxypyrrolidin-2-one 97%
(R)-4-Hydroxypyrrolidin-2-one


Formula: C₄H₇NO₂
Melting Pt: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00273364
CAS Number: 22677-21-0
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Specific optical rotation +54° to +60° (20°C, 589 nm) (c=0.36 in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty