Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44464-0010 44464-0050
TS44464-0010EA 126 USD InStock
TS44464-0010 TS44464-0050
4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester 97%
4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester


Formula: C₁₈H₂₈BNO₄
Melting Pt: 68…69 °C
Storage Temperature: Ambient
MDL Number: MFCD06409942
CAS Number: 330794-35-9
Order Now
Specification Test Results
Appearance Colorless to white to off-white Solid
Infrared spectrum Conforms
Melting point 66 to 71 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty