Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrophenylboronic acid pinacol ester 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44521-0010 44521-0050
TS44521-0010EA 74 USD InStock
TS44521-0010 TS44521-0050
2-Nitrophenylboronic acid pinacol ester 97%
2-Nitrophenylboronic acid pinacol ester


Formula: C₁₂H₁₆BNO₄
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD02179447
CAS Number: 190788-59-1
Order Now
Specification Test Results
Appearance Light yellow to brown Solid
Infrared spectrum Conforms
Melting point 43 to 50 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty