Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-methylnicotinate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44925-0050 44925-0250
TS44925-0050EA 60 USD InStock
TS44925-0050 TS44925-0250
Methyl-2-methylnicotinate 97%
Methyl-2-methylnicotinate


Formula: C₈H₉NO₂
Boiling Pt: 102…104 °C (10 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00051961
CAS Number: 65719-09-7
Order Now
Specification Test Results
Appearance Colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5145 to 1.5185 (20°C, 589 nm)
Water ≤1.5 %
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty