Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-Epoxystyrene 94%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

44945-0050 44945-0250
TS44945-0050EA 156 USD InStock
TS44945-0050 TS44945-0250
(S)-(-)-Epoxystyrene 94%
(S)-(-)-Epoxystyrene


Formula: C₈H₈O
Boiling Pt: 192…194 °C
Melting Pt: –37 °C
Density: 1.051 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064310
CAS Number: 20780-54-5
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5360 (20°C, 589 nm)
Water ≤0.5 %
GC ≥92 %
Specific optical rotation -31° to -35° (20°C, 589 nm) (neat)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty