Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-3-methyl-1-phenyl-1H-pyrazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44960-0050 44960-0250
TS44960-0050EA 48 USD InStock
TS44960-0050 TS44960-0250
5-Amino-3-methyl-1-phenyl-1H-pyrazole 97%
5-Amino-3-methyl-1-phenyl-1H-pyrazole


Formula: C₁₀H₁₁N₃
Boiling Pt: 333 °C (760 mmHg)
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00020727
CAS Number: 1131-18-6
Order Now
Specification Test Results
Appearance White to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 114 to 117 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty