Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-tert-Butyl-2,4-xylenol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

46257-0100 46257-0500
TS46257-0100EA 46 USD InStock
TS46257-0100 TS46257-0500
6-tert-Butyl-2,4-xylenol 97%
6-tert-Butyl-2,4-xylenol


Formula: C₁₂H₁₈O
Boiling Pt: 249 °C
Melting Pt: 22…23 °C
Storage Temperature: Ambient
MDL Number: MFCD00002234
CAS Number: 1879-09-0
UN: 2811
ADR: 6.1,I
Order Now
Specification Test Results
Appearance White to light yellow Liquid or solid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty