Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-3-tolualdehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

18844-0050 18844-0250
TS18844-0050EA 47 USD InStock
TS18844-0050 TS18844-0250
α,α,α-Trifluoro-3-tolualdehyde 97%
α,α,α-Trifluoro-3-tolualdehyde


Formula: C₈H₅F₃O
Boiling Pt: 83…86 °C (30 mmHg)
Density: 1.3 g/cm³
Flash Pt: 68 °C
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1
Order Now
Specification Test Results
Appearance Clear colorless to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4639 to 1.4659 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty