Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-3-iodobenzene 99% stabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20448-0050 20448-0250
TS20448-0050EA 31 USD InStock
TS20448-0050 TS20448-0250
1-Fluoro-3-iodobenzene 99% stabilised
1-Fluoro-3-iodobenzene


Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 183
Melting Pt: -42
Density: 1.89 g/cm³
Flash Pt: 67 °C
MDL Number: MFCD00001044
CAS Number: 1121-86-4
EINECS: 214-339-9
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5830 to 1.5860 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty