Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Fluoro-4-iodobenzene 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20450-0010 20450-0050 20450-0500
TS20450-0010EA 18 USD InStock
TS20450-0010 TS20450-0050 TS20450-0500
1-Fluoro-4-iodobenzene 99%
1-Fluoro-4-iodobenzene


Formula: C₆H₄FI
Boiling Pt: 182…184 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.925 g/cm³
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001052
CAS Number: 352-34-1
EINECS: 206-522-7
Order Now
Specification Test Results
Appearance Clear yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5840 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty