Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dibromobiphenyl (PBB No. 15) 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

21208-0250 21208-1000 21208-5000
TS21208-0250EA 29 USD InStock
TS21208-0250 TS21208-1000 TS21208-5000
4,4'-Dibromobiphenyl (PBB No. 15) 99%
4,4'-Dibromobiphenyl (PBB No. 15)


Formula: C₁₂H₈Br₂
MW: 312 g/mol
Boiling Pt: 357
Melting Pt: 164
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000101
CAS Number: 92-86-4
EINECS: 202-198-6
UN: 3152
ADR: 9 II
Order Now
Specification Test Results
Appearance White to yellow to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 165 to 169 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty