Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Bis(trifluoromethyl)benzoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

21446-0010 21446-0050
TS21446-0010EA 33 USD InStock
TS21446-0010 TS21446-0050
2,6-Bis(trifluoromethyl)benzoic acid 98%
2,6-Bis(trifluoromethyl)benzoic acid


Formula: C₉H₄F₆O₂
MW: 258.12
Boiling Pt: 260
Melting Pt: 137.7…138.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00000376
CAS Number: 24821-22-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty