Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl vinyl sulphone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22125-0010 22125-0050 22125-0250
TS22125-0010EA 36 USD InStock
TS22125-0010 TS22125-0050 TS22125-0250
Phenyl vinyl sulphone 98%
Phenyl vinyl sulphone


Formula: C₈H₈O₂S
MW: 168.22 g/mol
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00007554
CAS Number: 5535-48-8
Order Now
Specification Test Results
Appearance White to beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 66 to 70 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty