Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-α-Methoxyphenylacetic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

22447-0010 22447-0050
TS22447-0010EA 108 USD InStock
TS22447-0010 TS22447-0050
(R)-(-)-α-Methoxyphenylacetic acid 99%
(R)-(-)-α-Methoxyphenylacetic acid


Formula: C₉H₁₀O₃
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00004250
CAS Number: 3966-32-3
EINECS: 223-580-9
Order Now
Specification Test Results
Appearance Colorless to white Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 63 to 68 °C
Specific optical rotation -158° to -162° (20 °C, 589 nm) (c=1, MeOH)
Titration with NaOH 98.5 to 101.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty