Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-nitrophenol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

24197-0250 24197-1000
TS24197-0250EA 61 USD InStock
TS24197-0250 TS24197-1000
5-Fluoro-2-nitrophenol 99%
5-Fluoro-2-nitrophenol


Formula: C₆H₄FNO₃
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 32…35 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007107
CAS Number: 446-36-6
EINECS: 207-168-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow to orange-brown Powder or crystals or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 32 to 35 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty