Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,4-difluorobenzene 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

24936-0100 24936-0500
TS24936-0100EA 28 USD InStock
TS24936-0100 TS24936-0500
1-Bromo-2,4-difluorobenzene 98+%
1-Bromo-2,4-difluorobenzene


Formula: C₆H₃BrF₂
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.708 g/cm³
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000330
CAS Number: 348-57-2
EINECS: 206-479-4
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5060 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty