Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-iodobenzene 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

24942-0100 24942-0500
TS24942-0100EA 56 USD InStock
TS24942-0100 TS24942-0500
1-Bromo-3-iodobenzene 98+%
1-Bromo-3-iodobenzene


Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 240
Melting Pt: -9.3
Density: 2.21 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6590 to 1.6620 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty