Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-iodobenzoate 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

24983-0250 24983-1000
TS24983-0250EA 98 USD InStock
TS24983-0250 TS24983-1000
Methyl-2-iodobenzoate 99+%
Methyl-2-iodobenzoate


Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 272
Density: 1.73 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00016351
CAS Number: 610-97-9
EINECS: 210-243-6
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6020 to 1.6060 (20°C, 589 nm)
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty