Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorobenzonitrile 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

25005-0050 25005-0250 25005-1000
TS25005-0050EA 28 USD InStock
TS25005-0050 TS25005-0250 TS25005-1000
2-Fluorobenzonitrile 99%
2-Fluorobenzonitrile


Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 190
Melting Pt: 35…37 °C
Density: 1.11 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00001773
CAS Number: 394-47-8
EINECS: 206-897-7
UN: 3276
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5063 to 1.5083 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty