Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylethyl isothiocyanate 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

26516-0050 26516-0250
TS26516-0050EA 40 USD InStock
TS26516-0050 TS26516-0250
2-Phenylethyl isothiocyanate 96%
2-Phenylethyl isothiocyanate


Formula: C₆H₅CH₂CH₂NCS
Boiling Pt: 113 °C (1 mmHg)
Density: 1.1 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004821
CAS Number: 2257-09-2
EINECS: 218-855-5
UN: 2810
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5860 to 1.5910 (20°C, 589 nm)
GC ≥95 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty