Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Difluorobenzylamine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30566-0010 30566-0050 30566-0250
TS30566-0010EA 14 USD InStock
TS30566-0010 TS30566-0050 TS30566-0250
2,4-Difluorobenzylamine 98%
2,4-Difluorobenzylamine


Formula: C₇H₇F₂N
Boiling Pt: 82…84 °C (15 mmHg)
Density: 1.2 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00010142
CAS Number: 72235-52-0
EINECS: 276-502-0
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4930 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty