Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(Hexafluoroisopropylidene)dianiline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

31099-0010 31099-0050
TS31099-0010EA 56 USD InStock
TS31099-0010 TS31099-0050
4,4'-(Hexafluoroisopropylidene)dianiline 98%
4,4'-(Hexafluoroisopropylidene)dianiline


Formula: C₁₅H₁₂F₆N₂
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00039146
CAS Number: 1095-78-9
Order Now
Specification Test Results
Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 195 to 199 °C
Titration with HClO4 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty