Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzoyl isothiocyanate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

31463-0050 31463-0500
TS31463-0050EA 49 USD InStock
TS31463-0050 TS31463-0500
Benzoyl isothiocyanate 98%
Benzoyl isothiocyanate


Formula: C₈H₅NOS
MW: 163.2 g/mol
Boiling Pt: 252
Melting Pt: -1
Density: 1.21 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004815
CAS Number: 532-55-8
EINECS: 208-540-0
UN: 2810
ADR: 6.1 III
Merck Index: 13,01115
Order Now
Specification Test Results
Appearance Clear yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6350 to 1.6385 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty