Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-3,4-dichlorotoluene 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

32616-0250 32616-1000
TS32616-0250EA 75 USD InStock
TS32616-0250 TS32616-1000 TS32616-5000
α-Bromo-3,4-dichlorotoluene 97%
α-Bromo-3,4-dichlorotoluene


Formula: C₇H₅BrCl₂
Boiling Pt: 115…120 °C (1 mmHg)
Density: 1.65 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00041363
CAS Number: 18880-04-1
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6050 to 1.6080 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty