Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-o-phenylenediamine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33186-0050 33186-0250 33186-1000
TS33186-0050EA 27 USD InStock
TS33186-0050 TS33186-0250 TS33186-1000
4-Chloro-o-phenylenediamine 97%
4-Chloro-o-phenylenediamine


Formula: C₆H₇ClN₂
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 70…75 °C
MDL Number: MFCD00011691
CAS Number: 95-83-0
EINECS: 202-456-8
Order Now
Specification Test Results
Appearance Grey-brown to dark brown Powder or granules and/or chunks
Infrared spectrum Conforms
Melting point 70 to 75 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty