Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-bromobenzoate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33365-0050 33365-0250
TS33365-0050EA 37 USD InStock
TS33365-0050 TS33365-0250
Methyl-3-bromobenzoate 98%
Methyl-3-bromobenzoate


Formula: C₈H₇BrO₂
Boiling Pt: 139…140 °C (35 mmHg)
Melting Pt: 31 °C
MDL Number: MFCD00017777
CAS Number: 618-89-3
EINECS: 210-569-9
Order Now
Specification Test Results
Appearance White to light yellow Low melting crystalline solid or liquid
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty