Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-2,4,6-tribromobenzoic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33489-0050 33489-0250
TS33489-0050EA 48 USD InStock
TS33489-0050 TS33489-0250
3-Hydroxy-2,4,6-tribromobenzoic acid 97%
3-Hydroxy-2,4,6-tribromobenzoic acid


Formula: C₇H₃Br₃O₃
MW: 374.81 g/mol
Boiling Pt: 396
Melting Pt: 146
MDL Number: MFCD00055557
CAS Number: 14348-40-4
UN: 2811
Order Now
Specification Test Results
Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 144 to 149 °C
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty