Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34873-0010 34873-0050
TS34873-0010EA 413 USD InStock
TS34873-0010 TS34873-0050
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol 97%
(3S,4S)-(+)-1-Benzyl-3,4-pyrrolidindiol


Formula: C₁₁H₁₅NO₂
Melting Pt: 95…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01073893
CAS Number: 90365-74-5
Order Now
Specification Test Results
Appearance Off-white to yellow to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 95 to 101 °C
GC ≥96.0 %
Specific optical rotation +29° to +35° (20 °C, 589 nm) (c=1, MeOH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty