Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dibromobenzene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34893-0250 34893-1000
TS34893-0250EA 87 USD InStock
TS34893-0250 TS34893-1000
1,2-Dibromobenzene 98%
1,2-Dibromobenzene


Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 224
Melting Pt: 4…6 °C
Density: 1.956 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000057
CAS Number: 583-53-9
EINECS: 209-507-3
UN: 3082
ADR: 9 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.6100 to 1.6120 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty