Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Bromophenyl)acetone 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

35059-0010 35059-0050
TS35059-0010EA 15 USD InStock
TS35059-0010 TS35059-0050
(2-Bromophenyl)acetone 99%
(2-Bromophenyl)acetone


Formula: C₉H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD03410431
CAS Number: 21906-31-0
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5595 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty