Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromophenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35894-0010 35894-0050
TS35894-0010EA 32 USD InStock
TS35894-0010 TS35894-0050
3-Bromophenylboronic acid 97%
3-Bromophenylboronic acid


Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Melting Pt: 175…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00239386
CAS Number: 89598-96-9
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty