Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorophenylboronic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35934-0010 35934-0050
TS35934-0010EA 48 USD InStock
TS35934-0010 TS35934-0050
2-Fluorophenylboronic acid 98%
2-Fluorophenylboronic acid


Formula: C₆H₆BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00674013
CAS Number: 1993-03-9
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty