Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Formylphenylboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35937-0010 35937-0050 35937-0250
TS35937-0010EA 31 USD InStock
TS35937-0010 TS35937-0050 TS35937-0250
3-Formylphenylboronic acid 97%
3-Formylphenylboronic acid


Formula: C₇H₇BO₃
MW: 149.94
Melting Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00161356
CAS Number: 87199-16-4
Order Now
Specification Test Results
Appearance Off-white to beige to light orange Powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty