Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)phenylboronic acid 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35975-0010 35975-0050
TS35975-0010EA 38 USD InStock
TS35975-0010 TS35975-0050
3-(Trifluoromethyl)phenylboronic acid 97+%
3-(Trifluoromethyl)phenylboronic acid


Formula: C₇H₆BF₃O₂
MW: 189.93 g/mol
Melting Pt: 160…165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151854
CAS Number: 1423-26-3
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 161 to 167 °C
HPLC ≥97.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty