Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylresorcinol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

36474-0250 36474-1000
TS36474-0250EA 50 USD InStock
TS36474-0250 TS36474-1000
2-Methylresorcinol 98%
2-Methylresorcinol


Formula: C₇H₈O₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 118…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00002271
CAS Number: 608-25-3
EINECS: 210-155-8
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige to pink to brown Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 124 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty