Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Isopropyl-2-methylphenol

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36834-0250 36834-1000
TS36834-0250EA 32 USD InStock
TS36834-0250 TS36834-1000
5-Isopropyl-2-methylphenol
5-Isopropyl-2-methylphenol


Formula: C₁₀H₁₄O
Boiling Pt: 238 °C (760 mmHg)
Melting Pt: 3 °C
Density: 0.976
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00002236
CAS Number: 499-75-2
UN: 3145
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5240 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty