Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl thiosalicylate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36960-0050 36960-0250
TS36960-0050EA 82 USD InStock
TS36960-0050 TS36960-0250
Methyl thiosalicylate 97%
Methyl thiosalicylate


Formula: C₈H₈O₂S
MW: 168.21 g/mol
Boiling Pt: 98…100 °C (2 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060678
CAS Number: 4892-02-8
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light green Liquid
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5950 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty