Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mexiletine hydrochloride

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

37291-0050 37291-0250 37291-1000
TS37291-0050EA 33 USD InStock
TS37291-0050 TS37291-0250 TS37291-1000
Mexiletine hydrochloride
Mexiletine hydrochloride


Formula: C₁₁H₁₇NO
Melting Pt: 202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216024
CAS Number: 5370-01-4
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 200 to 205 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty