Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromophenyl)acetone 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

38022-0050 38022-0250
TS38022-0050EA 75 USD InStock
TS38022-0050 TS38022-0250
(4-Bromophenyl)acetone 97%
(4-Bromophenyl)acetone


Formula: C₉H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD00210401
CAS Number: 6186-22-7
Order Now
Specification Test Results
Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5560 to 1.5590 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty