Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-m-toluic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38197-0010 38197-0050
TS38197-0010EA 71 USD InStock
TS38197-0010 TS38197-0050
6-Chloro-m-toluic acid 98%
6-Chloro-m-toluic acid


Formula: C₈H₇ClO₂
MW: 170.6
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00045798
CAS Number: 6342-60-5
Order Now
Specification Test Results
Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 148 to 151 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty