Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Di-tert-butylsalicylic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38686-0050 38686-0250
TS38686-0050EA 33 USD InStock
TS38686-0050 TS38686-0250
3,5-Di-tert-butylsalicylic acid 99%
3,5-Di-tert-butylsalicylic acid


Formula: C₁₅H₂₂O₃
Melting Pt: 158…162 °C
Flash Pt: 179 °C
Storage Temperature: Ambient
MDL Number: MFCD00059058
CAS Number: 19715-19-6
Order Now
Specification Test Results
Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 160 to 165 °C
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty