Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Diphenyl-2-propyn-1-ol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

38997-0050 38997-0250
TS38997-0050EA 43 USD InStock
TS38997-0050 TS38997-0250
1,1-Diphenyl-2-propyn-1-ol 98%
1,1-Diphenyl-2-propyn-1-ol


Formula: C₁₅H₁₂O
Boiling Pt: 183 °C (20 mmHg)
Melting Pt: 46…50 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00041570
CAS Number: 3923-52-2
Order Now
Specification Test Results
Appearance White to cream to light yellow Powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 44 to 50 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty