Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromobenzaldehyde ethylene acetal 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39324-0050 39324-0250
TS39324-0050EA 54 USD InStock
TS39324-0050 TS39324-0250
2-Bromobenzaldehyde ethylene acetal 98%
2-Bromobenzaldehyde ethylene acetal


Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00155124
CAS Number: 34824-58-3
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5670 to 1.5700 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty