Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-(methoxymethyl)benzene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39340-0010 39340-0050 39340-0250
TS39340-0010EA 38 USD InStock
TS39340-0010 TS39340-0050 TS39340-0250
1-Bromo-2-(methoxymethyl)benzene 98%
1-Bromo-2-(methoxymethyl)benzene


Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD03790887
CAS Number: 52711-30-5
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5470 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty