Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyloxyphenylhydrazine hydrochloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39402-0010 39402-0050
TS39402-0010EA 69 USD InStock
TS39402-0010 TS39402-0050 TS39402-2500
3-Benzyloxyphenylhydrazine hydrochloride 98%
3-Benzyloxyphenylhydrazine hydrochloride


Formula: C₁₃H₁₅ClN₂O
Melting Pt: 155…157 °C
Storage Temperature: Ambient
CAS Number: 59146-68-8
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Violet Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty